Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184825.1 | NP_001171754.1 | 2 | 354 | ||
NM_001184826.1 | NP_001171755.1 | 2 | 354 | |||
NM_001297773.1 | NP_001284702.1 | 2 | 302 | |||
NM_006905.2 | NP_008836.2 | 2 | 354 | |||
XM_005259065.3 | XP_005259122.1 | 2 | 353 | |||
XM_011527125.1 | XP_011525427.1 | 2 | 353 | |||
XM_017026994.1 | XP_016882483.1 | 2 | 353 | |||
GenBank mRNA | AK300152.1 | - | 2 | 309 | ||
AK311491.1 | - | 2 | 315 | |||
BC022338.1 | - | 2 | 354 | 4732357 | ||
BC058285.1 | - | 2 | 278 | 4770394 | ||
CR450329.1 | - | 2 | 216 | |||
J04539.1 | - | 2 | 276 | |||
M17908.1 | - | 2 | 282 | |||
M20879.1 | - | 2 | 302 | |||
M20881.1 | - | 2 | 333 | |||
M21822.1 | - | 2 | 289 | |||
M25385.1 | - | 2 | 252 | |||
M33663.1 | - | 2 | 279 | |||
M33664.1 | - | 2 | 216 | |||
M34715.1 | - | 2 | 377 | |||
M37397.1 | - | 2 | 306 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008405.1 | - | 2 | 313 | 4043484 |