Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 5 | 1468 | ||
NM_001316339.1 | NP_001303268.1 | 5 | 1381 | |||
NM_002780.4 | NP_002771.2 | 6 | 1747 | |||
NM_213633.2 | NP_998798.1 | 5 | 1468 | |||
XM_017026996.1 | XP_016882485.1 | 7 | 1809 | |||
XM_017027000.1 | XP_016882489.1 | 6 | 1534 | |||
GenBank mRNA | AF289606.1 | - | 6 | 1088 | ||
BC008405.1 | - | 8 | 1606 | 4043484 | ||
BC063127.1 | - | 6 | 1615 | 4768860 | ||
M33665.1 | - | 4 | 888 | |||
M94891.1 | - | 6 | 1595 | |||
U18468.1 | - | 6 | 1514 | |||
U18469.1 | - | 5 | 1257 | |||
X17097.1 | - | 6 | 1601 |