Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079684.2 | NP_001073152.1 | 4, 5 | 612 | ||
NM_001079685.2 | NP_001073153.1 | 4, 5 | 591 | |||
NM_001287335.1 | NP_001274264.1 | 2, 3 | 325 | |||
NM_001287336.1 | NP_001274265.1 | 3, 4 | 504 | |||
NM_020444.4 | NP_065177.2 | 5, 6 | 699 | |||
XM_005265941.1 | XP_005265998.1 | 4, 5 | 512 | |||
XM_005265946.4 | XP_005266003.1 | 4, 5 | 607 | |||
XM_011534595.1 | XP_011532897.1 | 4, 5 | 474 | |||
XM_011534596.1 | XP_011532898.1 | 3, 4 | 423 | |||
XM_017009651.1 | XP_016865140.1 | 3, 4 | 408 | |||
GenBank mRNA | AK315428.1 | - | 5, 6 | 656 | ||
AL390217.1 | - | 4, 5 | 570 | |||
BC010448.1 | - | 5, 6 | 634 | 4155679 | ||
BC017097.1 | - | 5, 6 | 656 | 3529175 | ||
BX648859.1 | - | 5, 6 | 698 | |||
CR457331.1 | - | 3, 4 | 326 | |||
DQ073392.1 | - | 3, 4 | 326 | |||
HY114499.1 | - | 2, 3 | 283 | |||
HY119081.1 | - | 504 |