Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305141.1 | NP_001292070.1 | 6 | 1012 | ||
NM_001305142.1 | NP_001292071.1 | 6 | 999 | |||
NM_020677.4 | NP_065728.1 | 6 | 1113 | |||
XM_006720905.3 | XP_006720968.1 | 7 | 1050 | |||
XM_006720906.2 | XP_006720969.1 | 7 | 1005 | |||
XM_017023485.1 | XP_016878974.1 | 5 | 976 | |||
XM_017023489.1 | XP_016878978.1 | 7 | 1101 | |||
XM_017023490.1 | XP_016878979.1 | 6 | 926 | |||
XM_017023491.1 | XP_016878980.1 | 4 | 788 | |||
XM_017023492.1 | XP_016878981.1 | 5 | 897 | |||
GenBank mRNA | AF225419.1 | - | 6 | 1118 | ||
AK098784.1 | - | 6 | 973 | |||
BC002927.2 | - | 6 | 893 | 3954035 | ||
BC007364.2 | - | 6 | 955 | 3630659 |