Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145079.1 | NP_001138551.1 | 16 | 1753 | ||
NM_020751.2 | NP_065802.1 | 16 | 1753 | |||
NR_026745.1 | 17 | 1818 | ||||
XM_011535168.1 | XP_011533470.1 | 16 | 1703 | |||
GenBank mRNA | AB032960.1 | - | 16 | 1643 | ||
AF116827.1 | - | 17 | 1701 | |||
AK026638.1 | - | 15 | 1420 | |||
AK294443.1 | - | 16 | 1753 | |||
AK310953.1 | - | 2 | 244 | |||
BC051723.1 | - | 16 | 1703 | 5263056 | ||
CR627406.1 | - | 16 | 1696 |