Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110514.1 | NP_001103984.1 | 18 | 2479 | ||
XM_006723600.1 | XP_006723663.1 | 18 | 2274 | |||
XM_017027983.1 | XP_016883472.1 | 19 | 2371 | |||
XM_017027984.1 | XP_016883473.1 | 19 | 2477 | |||
XM_017027985.1 | XP_016883474.1 | 12 | 1633 | |||
GenBank mRNA | AB037863.1 | - | 18 | 2357 | ||
BC019106.2 | - | 4 | 970 | |||
BC037555.1 | - | 10 | 1579 | |||
BC054347.1 | - | 3 | 1133 | |||
BG750986.1 | - | 1 | 44 |