Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021178.4 | NP_067001.3 | 5 | 697 | ||
NM_182849.2 | NP_878269.1 | 4 | 497 | |||
NM_182852.3 | NP_878272.1 | 6 | 878 | |||
GenBank mRNA | AF216381.1 | - | 1 | 208 | ||
AK026233.1 | - | 5 | 664 | |||
AK225253.1 | - | 5 | 669 | |||
AK225451.1 | - | 5 | 664 | |||
AK226037.1 | - | 5 | 669 | |||
AK309183.1 | - | 5 | 666 | |||
AL161994.1 | - | 5 | 677 | |||
BC000369.2 | - | 4 | 639 | 2819346 | ||
BC001218.1 | - | 4 | 642 | 3528496 | ||
BC004435.1 | - | 4 | 639 | 2819346 | ||
BI771433.1 | - | 5 | 680 | |||
BI917081.1 | - | 5 | 688 | |||
BM741413.1 | - | 4 | 464 | |||
BX161423.1 | - | 5 | 857 | |||
BX161424.1 | - | 5 | 825 | |||
BX247978.1 | - | 5 | 805 | |||
BX334446.2 | - | 5 | 820 |