Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130033.1 | NP_001123505.1 | 6 | 835 | ||
NM_001130034.1 | NP_001123506.1 | 7 | 681 | |||
NM_001130035.1 | NP_001123507.1 | 6 | 576 | |||
NM_001130036.1 | NP_001123508.1 | 5 | 708 | |||
NM_021200.2 | NP_067023.1 | 7 | 940 | |||
XM_011545192.2 | XP_011543494.1 | 7 | 1514 | |||
XM_011545193.2 | XP_011543495.1 | 6 | 1409 | |||
GenBank mRNA | AB209716.1 | - | 6 | 488 | ||
AF081583.1 | - | 6 | 457 | |||
AF093249.1 | - | 6 | 460 | |||
AF100612.1 | - | 7 | 565 | |||
AF101054.1 | - | 6 | 831 | |||
AK054725.1 | - | 2 | 593 | |||
AK289620.1 | - | 6 | 488 | |||
AK289909.1 | - | 6 | 454 | |||
AK290109.1 | - | 5 | 708 | |||
AK301299.1 | - | 7 | 576 | |||
AK311158.1 | - | 7 | 333 | |||
AK314748.1 | - | 7 | 537 | |||
BC008075.2 | - | 6 | 514 | 3050369 | ||
U89715.1 | - | 7 | 936 |