Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286134.1 | NP_001273063.1 | 9 | 2736 | ||
NM_021932.5 | NP_068751.4 | 9 | 2754 | |||
XM_005253052.3 | XP_005253109.1 | 9 | 1922 | |||
XM_017018107.1 | XP_016873596.1 | 9 | 2006 | |||
GenBank mRNA | AK022870.1 | - | 8 | 1291 | ||
AK026897.1 | - | 9 | 2735 | |||
AK027090.1 | - | 6 | 663 | |||
AK027461.1 | - | 9 | 1424 | |||
AK225420.1 | - | 9 | 1422 | |||
AK314568.1 | - | 9 | 1443 | |||
AL136935.1 | - | 9 | 1735 | |||
AL390088.1 | - | 9 | 1457 | |||
BC011821.1 | - | 7 | 1200 | |||
BC111499.1 | - | 9 | 1550 | 6526180 | ||
BC121807.1 | - | 9 | 1630 | 40108083 | ||
BC121808.1 | - | 9 | 1630 | 40108080 | ||
BX538115.1 | - | 7 | 1598 |