Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 8 | 1024 | ||
NM_199053.2 | NP_951008.1 | 8 | 1024 | |||
XM_017008537.1 | XP_016864026.1 | 8 | 1024 | |||
XM_017008538.1 | XP_016864027.1 | 8 | 1024 | |||
XM_017008539.1 | XP_016864028.1 | 7 | 769 | |||
GenBank mRNA | AK091146.1 | - | 3 | 248 | ||
AK096345.1 | - | 8 | 998 | |||
AK315057.1 | - | 3 | 230 | |||
AL833451.1 | - | 7 | 832 | |||
AL833571.1 | - | 8 | 1003 | |||
BC051724.1 | - | 8 | 968 | 5265781 | ||
BX538044.1 | - | 8 | 978 | |||
BX647127.1 | - | 8 | 995 | |||
BX648499.1 | - | 8 | 999 |