Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000661.4 | NP_000652.2 | 5, 6 | 457 | ||
NM_001024921.3 | NP_001020092.1 | 5, 6 | 615 | |||
GenBank mRNA | AB062431.1 | - | 1 | 404 | ||
AK130966.1 | - | 1 | 420 | |||
AK312241.1 | - | 1 | 424 | |||
AY320412.1 | - | 5, 6 | 440 | |||
AY376242.1 | - | 4, 5 | 574 | |||
AY597415.1 | - | 1 | 431 | |||
BC000483.2 | - | 1 | 409 | 2964733 | ||
BC004156.2 | - | 1 | 403 | 2961026 | ||
BC004206.2 | - | 1 | 403 | 2961026 | ||
BC007967.2 | - | 1 | 459 | 4304670 | ||
BC012149.1 | - | 1 | 459 | 4554838 | ||
BC031906.1 | - | 4, 5 | 404 | 4996986 | ||
BC066318.1 | - | 1 | 424 | 5277491 | ||
BC070214.1 | - | 1 | 463 | 6605055 | ||
BC100285.1 | - | 4, 5 | 564 | 6022858 | ||
BG829769.1 | - | 4, 5 | 405 | |||
D14531.1 | - | 4, 5 | 404 | |||
U09953.1 | - | 5, 6 | 427 | |||
U21138.1 | - | 1 | 404 |
Gene Symbol: RPL9P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130277.1 | - | 1 | 1033 |