Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 5 | 2208 | ||
NM_206852.2 | NP_996734.1 | 3 | 603 | |||
XM_017021578.1 | XP_016877067.1 | 3 | 492 | |||
GenBank mRNA | AK128113.1 | - | 1 | 622 | ||
AK290534.1 | - | 3 | 603 | |||
AK297492.1 | - | 4 | 510 | |||
BC000314.2 | - | 3 | 388 | |||
BC003003.1 | - | 3 | 388 | |||
BC090862.1 | - | 5 | 2123 | 6179803 | ||
BC111694.1 | - | 6 | 1898 | 40035553 | ||
BX449967.2 | - | 3 | 591 | |||
CR457030.1 | - | 3 | 295 | |||
L10333.1 | - | 5 | 2121 | |||
L10334.1 | - | 3 | 1029 | |||
L10335.1 | - | 3 | 323 |