Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3GAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172435.1 | NP_001165906.1 | 11 | 981 | ||
NM_012233.2 | NP_036365.1 | 11 | 981 | |||
XM_011510823.2 | XP_011509125.1 | 11 | 981 | |||
XM_011510825.2 | XP_011509127.1 | 11 | 981 | |||
GenBank mRNA | AB449877.1 | - | 11 | 938 | ||
AK022494.1 | - | 11 | 953 | |||
BC146809.1 | - | 13 | 1218 | 8860368 | ||
D31886.1 | - | 11 | 940 |