Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035512.1 | NP_001030589.1 | 4 | 267 | ||
NM_001035513.1 | NP_001030590.1 | 3 | 210 | |||
NM_003001.3 | NP_002992.1 | 5 | 369 | |||
NR_103459.1 | 6 | 396 | ||||
GenBank mRNA | AB201252.1 | - | 3 | 198 | ||
AB211234.1 | - | 4 | 261 | |||
AK293687.1 | - | 4 | 259 | |||
BC020808.1 | - | 5 | 364 | 4291847 | ||
BC033626.1 | - | 5 | 362 | 5416956 | ||
BC066329.1 | - | 5 | 364 | 5301907 | ||
BG724095.1 | - | 5 | 493 | |||
CR457102.1 | - | 4 | 339 | |||
D49737.1 | - | 5 | 352 | |||
DA751501.1 | - | 6 | 381 | |||
KU178321.1 | - | 5 | 339 | |||
KU178322.1 | - | 2 | 180 | |||
U57877.1 | - | 5 | 366 |