Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256410.1 | NP_001243339.1 | 2 | 284 | ||
NM_001256411.1 | NP_001243340.1 | 2 | 284 | |||
NM_001256412.1 | NP_001243341.1 | 2 | 284 | |||
NM_001256415.1 | NP_001243344.1 | 2 | 284 | |||
NM_021252.4 | NP_067075.1 | 2 | 284 | |||
NR_046172.1 | 2 | 284 | ||||
GenBank mRNA | AF087860.1 | - | 2 | 142 | ||
AF136974.1 | - | 2 | 198 | |||
AF137372.1 | - | 2 | 88 | |||
AF498950.1 | - | 2 | 88 | |||
AK001555.1 | - | 2 | 88 | |||
AK223153.1 | - | 2 | 111 | |||
AK295443.1 | - | 2 | 154 | |||
AK297629.1 | - | 2 | 154 | |||
AK299457.1 | - | 2 | 154 | |||
AL136734.1 | - | 2 | 137 | |||
AY574034.1 | - | 2 | 173 | |||
BC015014.2 | - | 2 | 138 | 3922029 | ||
BC029350.1 | - | 2 | 154 | 4819866 | ||
BP210053.1 | - | 2 | 148 | |||
BT009840.1 | - | 2 | 88 | |||
CN357871.1 | - | 2 | 353 | |||
CR533455.1 | - | 2 | 88 |