Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022451.10 | NP_071896.8 | 7 | 858 | ||
XM_017016544.1 | XP_016872033.1 | 5 | 911 | |||
GenBank mRNA | AB077992.1 | - | 7 | 745 | ||
AK091246.1 | - | 7 | 815 | |||
AK292520.1 | - | 7 | 840 | |||
BC049850.1 | - | 7 | 808 | 5549517 |