Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018452.5 | NP_060922.2 | 1, 2 | 848 | ||
GenBank mRNA | AF161405.1 | - | 1, 2 | 185 | ||
AF217510.1 | - | 1, 2 | 190 | |||
AK000891.1 | - | 1, 2 | 96 | |||
BC029130.1 | - | 1, 2 | 116 | 5182086 | ||
BC146893.1 | - | 1, 2 | 158 | 9056821 | ||
BC146895.1 | - | 1, 2 | 158 | 9056823 | ||
DA684766.1 | - | 1, 2 | 351 |