Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022570.4 | NP_072092.2 | 3 | 480 | ||
NM_197947.2 | NP_922938.1 | 4 | 618 | |||
NM_197948.2 | NP_922939.1 | 4 | 618 | |||
NM_197949.2 | NP_922940.1 | 3 | 480 | |||
NM_197950.2 | NP_922941.1 | 2 | 381 | |||
NR_125336.1 | 4 | 618 | ||||
XM_006719135.3 | XP_006719198.1 | 4 | 613 | |||
XM_017019822.1 | XP_016875311.1 | 4 | 612 | |||
XM_017019823.1 | XP_016875312.1 | 3 | 377 | |||
GenBank mRNA | AF313468.1 | - | 5 | 482 | ||
AF313469.1 | - | 3 | 343 | |||
AF400595.1 | - | 4 | 431 | |||
AF400596.1 | - | 3 | 293 | |||
AF400597.1 | - | 4 | 431 | |||
AF400598.1 | - | 3 | 293 | |||
AF400599.1 | - | 2 | 194 | |||
AF400601.1 | - | 4 | 431 | |||
AF400602.1 | - | 5 | 481 | |||
AJ312372.1 | - | 3 | 448 | |||
AJ312373.1 | - | 4 | 586 | |||
AK294111.1 | - | 4 | 610 | |||
AK297028.1 | - | 3 | 364 | |||
AK298679.1 | - | 4 | 534 | |||
AK298724.1 | - | 2 | 297 | |||
AK313247.1 | - | 3 | 411 | |||
AY009090.2 | - | 3 | 364 | |||
AY026769.2 | - | 4 | 431 | |||
AY026770.2 | - | 3 | 293 | |||
AY026771.1 | - | 3 | 293 | |||
AY359002.1 | - | 3 | 452 | |||
BC071746.1 | - | 3 | 394 | 4690475 | ||
CR749536.1 | - | 3 | 1600 |