Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000994.3 | NP_000985.1 | 3 | 230 | ||
NM_001007073.1 | NP_001007074.1 | 3 | 329 | |||
NM_001007074.1 | NP_001007075.1 | 2 | 349 | |||
GenBank mRNA | AK311909.1 | - | 3 | 187 | ||
BC011514.1 | - | 1 | 156 | 2900089 | ||
BC070209.1 | - | 1 | 162 | 6738371 | ||
BQ068496.1 | - | 267 | ||||
CF124158.1 | - | 3 | 267 | |||
X03342.1 | - | 3 | 170 |