Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012311.3 | NP_036443.1 | 2 | 228 | ||
NR_045609.1 | 2 | 228 | ||||
XM_006717434.3 | XP_006717497.1 | 2 | 223 | |||
XM_011519427.1 | XP_011517729.1 | 2 | 223 | |||
GenBank mRNA | AJ005273.1 | - | 2 | 199 | ||
AK301789.1 | - | 2 | 228 | |||
AK309059.1 | - | 2 | 164 | |||
BC017309.2 | - | 2 | 145 | 5089578 | ||
BX504040.1 | - | 2 | 195 |