Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 20 | 4302 | ||
XM_006718795.3 | XP_006718858.1 | 9 | 2816 | |||
XM_011542691.2 | XP_011540993.1 | 21 | 4989 | |||
XM_011542692.2 | XP_011540994.1 | 21 | 4977 | |||
XM_011542695.2 | XP_011540997.1 | 20 | 4520 | |||
XM_017017393.1 | XP_016872882.1 | 20 | 5007 | |||
XM_017017396.1 | XP_016872885.1 | 14 | 4331 |