Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012311.3 | NP_036443.1 | 8 | 840 | ||
NR_045609.1 | 8 | 840 | ||||
XM_006717434.3 | XP_006717497.1 | 8 | 835 | |||
XM_006717435.3 | XP_006717498.1 | 7 | 708 | |||
XM_011519427.1 | XP_011517729.1 | 8 | 835 | |||
XM_017015946.1 | XP_016871435.1 | 7 | 711 | |||
GenBank mRNA | AJ005273.1 | - | 8 | 811 | ||
AK296637.1 | - | 7 | 708 | |||
AK301789.1 | - | 8 | 840 | |||
AK309059.1 | - | 8 | 776 | |||
BC017309.2 | - | 8 | 757 | 5089578 |