Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320327.1 | NP_001307256.1 | 3 | 624 | ||
NM_016139.3 | NP_057223.1 | 3 | 624 | |||
GenBank mRNA | AF078845.1 | - | 3 | 488 | ||
AY605046.1 | - | 3 | 407 | |||
AY633613.1 | - | 3 | 428 | |||
BC003079.2 | - | 3 | 444 | 2988156 | ||
BC015639.2 | - | 3 | 428 | 4637353 | ||
BC066331.1 | - | 3 | 447 | 4792941 | ||
BC071985.1 | - | 3 | 444 | 6426679 | ||
BC100275.1 | - | 3 | 440 | 6192342 | ||
BF347122.1 | - | 498 | ||||
CN363208.1 | - | 3 | 464 |