Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024505.1 | 4 | 458 | |||
NR_024506.1 | 4 | 458 | ||||
NR_024507.2 | 4 | 458 | ||||
GenBank mRNA | AY116214.1 | - | 4 | 458 |