Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032478.3 | NP_115867.2 | 8 | 1201 | ||
GenBank mRNA | AF161380.1 | - | 4 | 360 | ||
AK313283.1 | - | 8 | 1016 | |||
BC013311.2 | - | 8 | 1000 | 3546083 | ||
BC034460.1 | - | 8 | 955 |