Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004536.2 | NP_004527.2 | 15 | 4363 | ||
NM_022892.1 | NP_075043.1 | 13 | 3692 | |||
XM_011543413.2 | XP_011541715.1 | 5 | 2501 | |||
XM_017009490.1 | XP_016864979.1 | 5 | 3048 | |||
GenBank mRNA | AB048534.1 | - | 13 | 3692 | ||
AB209123.1 | - | 4 | 3749 | |||
AK124511.1 | - | 4 | 3044 | |||
AK295115.1 | - | 4 | 1591 | |||
BC136273.1 | - | 13 | 3700 | 9020260 | ||
BC143761.1 | - | 12 | 3532 | 9052275 | ||
U19251.1 | - | 15 | 3936 |