Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003133.5 | NP_003124.1 | 2, 3 | 264 | ||
GenBank mRNA | AF070649.1 | - | 2, 3 | 243 | ||
AK289595.1 | - | 2, 3 | 229 | |||
BC008443.1 | - | 2, 3 | 248 | 4102222 | ||
BC015094.1 | - | 1 | 235 | 3930047 | ||
BC017414.1 | - | 1 | 15 | |||
BC022415.1 | - | 1 | 230 | 4243562 | ||
BC066957.1 | - | 2, 3 | 251 | 5263635 | ||
U20998.1 | - | 2, 3 | 242 |