Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 6 | 1952 | ||
NM_001164644.1 | NP_001158116.1 | 6 | 1922 | |||
NM_001164645.1 | NP_001158117.1 | 6 | 1808 | |||
NM_001164646.1 | NP_001158118.1 | 6 | 1816 | |||
NM_001164647.1 | NP_001158119.1 | 5 | 1892 | |||
NM_005411.4 | NP_005402.3 | 6 | 1963 | |||
XM_005270062.4 | XP_005270119.1 | 5 | 1952 | |||
XM_006717953.2 | XP_006718016.1 | 6 | 1945 | |||
GenBank mRNA | BC026229.1 | - | 2 | 1435 | ||
HQ021433.2 | - | 5 | 1892 | |||
HQ021434.1 | - | 5 | 1903 | |||
HQ021435.1 | - | 5 | 1892 | |||
HQ021436.1 | - | 5 | 1891 | |||
HQ021437.1 | - | 6 | 1974 | |||
HQ021438.1 | - | 6 | 1963 | |||
HQ021439.1 | - | 6 | 1962 | |||
HQ021440.1 | - | 6 | 1963 | |||
HQ021441.1 | - | 6 | 1952 | |||
HQ021442.1 | - | 6 | 1951 | |||
JX502764.1 | - | 6 | 1963 | |||
JX502765.1 | - | 6 | 1952 |