Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR89B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016334.3 | NP_057418.1 | 4 | 436 | ||
XM_005277401.3 | XP_005277458.1 | 4 | 409 | |||
XM_005277402.4 | XP_005277459.1 | 4 | 573 | |||
XM_005277404.3 | XP_005277461.1 | 4 | 409 | |||
XM_006711377.3 | XP_006711440.1 | 5 | 472 | |||
XM_006711378.2 | XP_006711441.1 | 5 | 455 | |||
XM_011509610.2 | XP_011507912.1 | 5 | 469 | |||
XM_011509612.2 | XP_011507914.1 | 5 | 410 | |||
XM_011509613.2 | XP_011507915.1 | 4 | 409 | |||
XM_017001448.1 | XP_016856937.1 | 5 | 972 | |||
XM_017001449.1 | XP_016856938.1 | 4 | 409 | |||
GenBank mRNA | AK299995.1 | - | 4 | 407 | ||
AK302266.1 | - | 4 | 428 | |||
AL133020.1 | - | 4 | 420 | |||
DC305701.1 | - | 4 | 436 | |||
KJ534849.1 | - | 4 | 292 | |||
KJ534850.1 | - | 4 | 292 | |||
KJ534851.1 | - | 4 | 292 | |||
KJ535070.1 | - | 5 | 620 | |||
U78723.1 | - | 4 | 395 |
Gene Symbol: GPR89A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097612.1 | NP_001091081.1 | 4 | 466 | ||
NM_001097613.2 | NP_001091082.2 | 5 | 512 | |||
NR_036541.1 | 4 | 466 | ||||
XM_006711492.3 | XP_006711555.1 | 4 | 436 | |||
XM_006711493.3 | XP_006711556.1 | 5 | 424 | |||
XM_011509908.2 | XP_011508210.1 | 5 | 482 | |||
XM_011509909.2 | XP_011508211.1 | 4 | 694 | |||
XM_011509910.2 | XP_011508212.1 | 4 | 436 | |||
XM_017002150.1 | XP_016857639.1 | 4 | 436 | |||
GenBank mRNA | AB097024.1 | - | 4 | 407 | ||
AB097025.1 | - | 4 | 407 | |||
AF132947.1 | - | 4 | 391 | |||
AK021758.1 | - | 4 | 436 | |||
AK223223.1 | - | 4 | 407 | |||
AK297145.1 | - | 4 | 466 | |||
AK316371.1 | - | 5 | 424 | |||
AY358966.1 | - | 4 | 441 | |||
BC003187.1 | - | 4 | 369 | 3537949 | ||
BC067816.1 | - | 4 | 405 | 5295014 | ||
BC146880.1 | - | 4 | 387 | 9056808 | ||
BT006679.1 | - | 4 | 292 | |||
DC349534.1 | - | 4 | 466 | |||
KJ534847.1 | - | 4 | 292 | |||
KJ534848.1 | - | 4 | 292 | |||
KJ535029.1 | - | 4 | 292 | |||
KJ535049.1 | - | 4 | 292 |
Set Membership: |
G-protein coupled receptor |