Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM76A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143912.1 | NP_001137384.1 | 2 | 226 | ||
NM_001143913.1 | NP_001137385.1 | 2 | 226 | |||
NM_001143914.1 | NP_001137386.1 | 2 | 226 | |||
NM_001143915.1 | NP_001137387.1 | 2 | 226 | |||
NM_152660.2 | NP_689873.1 | 2 | 226 | |||
XM_017000543.1 | XP_016856032.1 | 2 | 177 | |||
GenBank mRNA | AK098318.1 | - | 2 | 177 | ||
AK290120.1 | - | 2 | 221 | |||
AK301672.1 | - | 2 | 209 | |||
AK310165.1 | - | 2 | 177 | |||
AL035307.1 | - | 2 | 148 | |||
AL035309.1 | - | 2 | 148 | |||
BC025768.1 | - | 2 | 226 | 5211010 |