Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 2 | 229 | ||
NM_001197114.1 | NP_001184043.1 | 2 | 229 | |||
GenBank mRNA | AK096479.1 | - | 2 | 135 | ||
DQ157847.1 | - | 1 | 48 | |||
DQ157848.1 | - | 1 | 48 |
Gene Symbol: IQCJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042705.2 | NP_001036170.1 | 2 | 229 | ||
NM_001042706.2 | NP_001036171.1 | 2 | 229 | |||
NM_001197100.1 | NP_001184029.1 | 2 | 229 | |||
GenBank mRNA | AK309049.1 | - | 2 | 153 | ||
BC137464.1 | - | 2 | 109 | 9021464 | ||
BC137469.1 | - | 2 | 109 | 9021469 | ||
BC144658.1 | - | 2 | 109 | 9053189 | ||
DA305468.1 | - | 2 | 233 | |||
DQ309553.1 | - | 2 | 153 | |||
DQ309554.1 | - | 2 | 153 |