Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s227452
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
4
1392
NM_001206856.2
NP_001193785.1
4
1392
NM_001319842.1
NP_001306771.1
4
1392
NM_001319843.1
NP_001306772.1
4
1392
NM_001319844.1
NP_001306773.1
5
647
NM_001319845.1
NP_001306774.1
5
647
NM_001319846.1
NP_001306775.1
5
637
NM_023925.4
NP_076414.2
4
1392
NM_032156.4
NP_115532.3
4
1392
XM_006719138.3
XP_006719201.1
4
1515
XM_006719139.3
XP_006719202.1
4
1515
XM_006719140.3
XP_006719203.1
4
1515
XM_006719142.3
XP_006719205.1
4
1515
XM_006719144.3
XP_006719207.1
4
1515
XM_006719145.3
XP_006719208.1
4
1515
XM_006719146.3
XP_006719209.1
4
1515
XM_006719147.3
XP_006719210.1
4
1515
XM_017019851.1
XP_016875340.1
4
1515
XM_017019852.1
XP_016875341.1
5
610
XM_017019853.1
XP_016875342.1
5
680
XM_017019854.1
XP_016875343.1
4
1515
XM_017019855.1
XP_016875344.1
5
637
XM_017019856.1
XP_016875345.1
5
637
XM_017019857.1
XP_016875346.1
5
610
XM_017019858.1
XP_016875347.1
5
637
XM_017019859.1
XP_016875348.1
5
610
XM_017019860.1
XP_016875349.1
4
1515
XM_017019861.1
XP_016875350.1
5
637
XM_017019862.1
XP_016875351.1
4
1515
XM_017019863.1
XP_016875352.1
4
1515
XM_017019864.1
XP_016875353.1
4
1515
XM_017019865.1
XP_016875354.1
4
1515
XM_017019866.1
XP_016875355.1
5
610
XM_017019867.1
XP_016875356.1
5
637
XM_017019868.1
XP_016875357.1
5
610
XM_017019869.1
XP_016875358.1
5
637
XM_017019870.1
XP_016875359.1
5
610
XM_017019871.1
XP_016875360.1
5
681
XM_017019872.1
XP_016875361.1
5
637
XM_017019873.1
XP_016875362.1
5
637
XM_017019874.1
XP_016875363.1
5
610
XM_017019875.1
XP_016875364.1
5
637
XM_017019876.1
XP_016875365.1
3
221
XM_017019877.1
XP_016875366.1
3
221
XM_017019878.1
XP_016875367.1
3
221
XM_017019879.1
XP_016875368.1
5
610
XM_017019880.1
XP_016875369.1
3
221
XM_017019881.1
XP_016875370.1
5
610
XM_017019882.1
XP_016875371.1
5
610
XM_017019883.1
XP_016875372.1
3
221
GenBank mRNA
AK021453.1
-
4
242
AY074490.1
-
4
1392
AY074491.1
-
4
1392
BC117672.1
-
4
1189
40068103
BC117673.1
-
4
1189
40068104
BX537569.1
-
4
541
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008763471.
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-gx66z:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline