Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272034.1 | NP_001258963.1 | 5 | 339 | ||
NM_006949.3 | NP_008880.2 | 4, 5 | 306 | |||
NR_073560.1 | 4, 5 | 306 | ||||
XM_011528210.1 | XP_011526512.1 | 4, 5 | 302 | |||
XM_011528212.2 | XP_011526514.1 | 4, 5 | 302 | |||
GenBank mRNA | AB002559.1 | - | 4, 5 | 281 | ||
AF318317.1 | - | 1 | 1477 | |||
AK222967.1 | - | 4, 5 | 283 | |||
AK222977.1 | - | 4, 5 | 283 | |||
AK302257.1 | - | 4, 5 | 261 | |||
AK303701.1 | - | 5 | 327 | |||
AK314230.1 | - | 4, 5 | 285 | |||
BQ067493.1 | - | 4, 5 | 307 | |||
DA431480.1 | - | 4, 5 | 302 | |||
U63533.1 | - | 4, 5 | 281 |