Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012067.2 | NP_036199.2 | 7 | 1213 | ||
GenBank mRNA | AF040639.1 | - | 7 | 1220 | ||
AJ271799.1 | - | 7 | 1143 | |||
BC025709.1 | - | 7 | 1199 | 5224626 | ||
BC031562.1 | - | 7 | 1155 | 5184829 | ||
BC042420.1 | - | 7 | 1179 | 5186889 | ||
BG747063.1 | - | 2 | 446 |