Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195545.1 | NP_001182474.1 | 2 | 108 | ||
XM_011524158.2 | XP_011522460.1 | 4 | 294 | |||
XM_017024003.1 | XP_016879492.1 | 4 | 273 | |||
GenBank mRNA | DY655553.1 | - | 2 | 108 |