Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077440.1 | NP_001070908.1 | 10 | 2487 | ||
NM_001077441.1 | NP_001070909.1 | 10 | 1974 | |||
NM_001301038.1 | NP_001287967.1 | 10 | 2487 | |||
NM_014739.2 | NP_055554.1 | 10 | 2493 | |||
XM_005267237.3 | XP_005267294.1 | 10 | 2429 | |||
XM_017011529.1 | XP_016867018.1 | 10 | 1961 | |||
XM_017011530.1 | XP_016867019.1 | 10 | 2768 | |||
XM_017011531.1 | XP_016867020.1 | 10 | 1904 | |||
XM_017011532.1 | XP_016867021.1 | 11 | 2656 | |||
GenBank mRNA | AF249273.1 | - | 8 | 2234 | ||
AK303559.1 | - | 1 | 353 | |||
AK308870.1 | - | 3 | 484 | |||
AK309259.1 | - | 3 | 707 | |||
AK315884.1 | - | 2 | 91 | |||
BC132780.1 | - | 10 | 2437 | 40146802 | ||
BC144281.1 | - | 10 | 2431 | 9052801 | ||
BC144299.1 | - | 11 | 2601 | |||
BX648431.1 | - | 2 | 155 | |||
BX648878.1 | - | 6 | 1234 | |||
BX649160.1 | - | 10 | 1905 | |||
D79986.1 | - | 10 | 2493 |