Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277864.1 | NP_001264793.1 | 5 | 933 | ||
NR_102687.1 | 4 | 668 | ||||
NR_102688.1 | 5 | 846 | ||||
NR_102689.1 | 5 | 610 | ||||
NR_102691.1 | 4 | 432 | ||||
NR_102692.1 | 5 | 693 | ||||
NR_102693.1 | 6 | 789 | ||||
NR_102694.1 | 4 | 558 | ||||
XM_006720246.3 | XP_006720309.1 | 7 | 661 | |||
GenBank mRNA | BC171914.1 | - | 5 | 652 | ||
BC171918.1 | - | 6 | 748 | |||
GQ871921.1 | - | 4 | 445 | |||
GQ871922.1 | - | 4 | 405 | |||
GQ871923.1 | - | 5 | 609 | |||
GQ871924.1 | - | 5 | 882 | |||
GQ871925.1 | - | 5 | 624 | |||
GQ871926.1 | - | 4 | 469 | |||
GQ871927.1 | - | 1 | 899 | |||
KU058702.1 | - | 5 | 870 | |||
KU058703.1 | - | 5 | 809 | |||
KU058704.1 | - | 5 | 787 | |||
KU058705.1 | - | 5 | 737 | |||
KU058706.1 | - | 4 | 735 | |||
KU058707.1 | - | 4 | 609 | |||
KU058708.1 | - | 5 | 591 | |||
KU058709.1 | - | 3 | 474 |