Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM178A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003370.1 | XP_016858859.1 | 1 | 65 | ||
GenBank mRNA | DA062982.1 | - | 1 | 170 |