Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109983.1 | 7, 8 | 881 | |||
GenBank mRNA | AK024248.1 | - | 7, 8 | 852 |
Gene Symbol: GTF2IP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132119.1 | 7, 8 | 1465 | |||
GenBank mRNA | CB122954.1 | - | 2, 3 | 112 |
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 5, 6 | 661 | |||
GenBank mRNA | AK293765.1 | - | 5, 6 | 661 |
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 2, 3 | 200 | |||
GenBank mRNA | AK302487.1 | - | 2, 3 | 206 | ||
AK304049.1 | - | 2, 3 | 180 | |||
BC062355.1 | - | 2, 3 | 104 | |||
BC131589.1 | - | 2, 3 | 104 |