Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176819.3 | NP_789789.2 | 5 | 1213 | ||
XM_005272670.1 | XP_005272727.1 | 4 | 1014 | |||
XM_006724559.1 | XP_006724622.1 | 4 | 936 | |||
GenBank mRNA | AK094877.1 | - | 3 | 438 | ||
AK294472.1 | - | 4 | 782 | |||
DA378381.1 | - | 2 | 307 |