Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014981.1 | NP_055796.1 | 21 | 2355 | ||
XM_011512559.2 | XP_011510861.1 | 22 | 2476 | |||
XM_017005922.1 | XP_016861411.1 | 12 | 1620 | |||
GenBank mRNA | AB023217.3 | - | 21 | 2355 | ||
AK126801.1 | - | 3 | 290 |