Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012308.2 | NP_036440.1 | 7 | 1439 | ||
NR_027473.1 | 7 | 1439 | ||||
XM_006718480.3 | XP_006718543.1 | 3 | 457 | |||
XM_011544860.2 | XP_011543162.1 | 7 | 1498 | |||
XM_011544861.1 | XP_011543163.1 | 2 | 176 | |||
XM_017017391.1 | XP_016872880.1 | 7 | 745 | |||
XM_017017392.1 | XP_016872881.1 | 6 | 556 | |||
GenBank mRNA | AB023221.2 | - | 7 | 756 | ||
AK024505.1 | - | 7 | 681 | |||
BC047371.1 | - | 2 | 176 | 5270744 | ||
BC047486.1 | - | 7 | 1441 | 5296445 | ||
BC064360.1 | - | 7 | 854 | 5534384 | ||
JQ710741.1 | - | 6 | 623 | |||
JQ710742.1 | - | 7 | 745 | |||
JQ710743.1 | - | 6 | 623 | |||
JQ710744.1 | - | 8 | 817 |