Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000718.3 | NP_000709.1 | 6, 7 | 1110 | ||
NM_001243812.1 | NP_001230741.1 | 6, 7 | 1110 | |||
XM_011518991.2 | XP_011517293.1 | 6, 7 | 1074 | |||
XM_011518992.2 | XP_011517294.1 | 6, 7 | 1074 | |||
XM_011518993.2 | XP_011517295.1 | 6, 7 | 1073 | |||
XM_011518994.2 | XP_011517296.1 | 6, 7 | 1073 | |||
XM_017015115.1 | XP_016870604.1 | 6, 7 | 1074 | |||
XM_017015116.1 | XP_016870605.1 | 6, 7 | 1074 | |||
XM_017015117.1 | XP_016870606.1 | 6, 7 | 1073 | |||
XM_017015118.1 | XP_016870607.1 | 6, 7 | 1073 | |||
XM_017015119.1 | XP_016870608.1 | 6, 7 | 1073 | |||
XM_017015120.1 | XP_016870609.1 | 6, 7 | 1073 | |||
GenBank mRNA | M94172.1 | - | 6, 7 | 1110 | ||
M94173.1 | - | 6, 7 | 1110 |