Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 10 | 924 | ||
NM_001144832.2 | NP_001138304.1 | 10 | 899 | |||
NM_001297662.1 | NP_001284591.1 | 10 | 1006 | |||
NM_001297663.1 | NP_001284592.1 | 10 | 901 | |||
NM_001297664.1 | NP_001284593.1 | 10 | 937 | |||
NM_001297665.1 | NP_001284594.1 | 10 | 1029 | |||
XM_005270644.2 | XP_005270701.1 | 10 | 930 | |||
XM_006710471.2 | XP_006710534.1 | 10 | 1027 | |||
XM_011541048.2 | XP_011539350.1 | 10 | 1004 | |||
XM_011541049.2 | XP_011539351.1 | 10 | 1143 | |||
XM_011541050.2 | XP_011539352.1 | 10 | 1004 | |||
XM_011541051.2 | XP_011539353.1 | 10 | 1037 | |||
XM_011541052.2 | XP_011539354.1 | 10 | 983 | |||
XM_011541053.2 | XP_011539355.1 | 10 | 1021 | |||
XM_011541054.2 | XP_011539356.1 | 10 | 857 | |||
XM_017000715.1 | XP_016856204.1 | 10 | 904 | |||
GenBank mRNA | AB007921.1 | - | 10 | 1052 | ||
AF007170.1 | - | 10 | 805 | |||
AK000220.1 | - | 10 | 1004 | |||
AK298019.1 | - | 10 | 908 | |||
AK301587.1 | - | 10 | 897 | |||
AK311430.1 | - | 10 | 826 | |||
BC028374.2 | - | 10 | 971 | |||
CX788693.1 | - | 10 | 839 |