Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277285.1 | NP_001264214.1 | 13, 14 | 1801 | ||
XM_006718795.3 | XP_006718858.1 | 2, 3 | 315 | |||
XM_011542691.2 | XP_011540993.1 | 14, 15 | 2488 | |||
XM_011542692.2 | XP_011540994.1 | 14, 15 | 2476 | |||
XM_011542694.2 | XP_011540996.1 | 14, 15 | 2491 | |||
XM_011542695.2 | XP_011540997.1 | 13, 14 | 2019 | |||
XM_017017393.1 | XP_016872882.1 | 13, 14 | 2506 | |||
XM_017017394.1 | XP_016872883.1 | 14, 15 | 2491 | |||
XM_017017395.1 | XP_016872884.1 | 14, 15 | 2490 | |||
XM_017017396.1 | XP_016872885.1 | 7, 8 | 1830 | |||
GenBank mRNA | AB028953.1 | - | 1, 2 | 43 | ||
AK097578.1 | - | 10, 11 | 1531 |
Set Membership: |
G-protein coupled receptor |