Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORD89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126261.1 | - | 1 | 1724 |
Gene Symbol: RNF149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK129799.1 | - | 8 | 1515 |