Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005781.1 | NP_001005781.1 | 3 | 288 | ||
NM_001005782.1 | NP_001005782.1 | 2 | 213 | |||
NM_003352.4 | NP_003343.1 | 3 | 288 | |||
GenBank mRNA | AB062294.1 | - | 3 | 264 | ||
AK311840.1 | - | 3 | 253 | |||
BC006462.1 | - | 3 | 271 | 3546125 | ||
BC053528.1 | - | 3 | 246 | 3452929 | ||
BC066306.1 | - | 3 | 264 | 4818792 | ||
BG569821.1 | - | 2 | 181 | |||
BT006632.1 | - | 1 | 140 | |||
BU532734.1 | - | 237 | ||||
CR542147.1 | - | 1 | 140 | |||
CR542156.1 | - | 1 | 140 | |||
U38784.1 | - | 4 | 206 | |||
U61397.1 | - | 3 | 285 | |||
U67122.1 | - | 3 | 220 | |||
U72722.1 | - | 1 | 140 | |||
U83117.1 | - | 3 | 275 | |||
X99586.1 | - | 181 |
Gene Symbol: SUMO1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002190.1 | 1 | 276 |