Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003395.2 | NP_003386.1 | 4 | 1122 | ||
XM_011544271.2 | XP_011542573.1 | 4 | 1605 | |||
GenBank mRNA | AB060283.1 | - | 4 | 1045 | ||
BC111960.1 | - | 4 | 1122 | 8327428 | ||
BC113431.1 | - | 4 | 1122 | 8322483 |