Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM106C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143842.1 | NP_001137314.1 | 5, 6 | 738 | ||
NM_024056.3 | NP_076961.1 | 5, 6 | 667 | |||
GenBank mRNA | AK310035.1 | - | 3, 4 | 1215 | ||
AK313698.1 | - | 5, 6 | 609 | |||
BC000854.2 | - | 4, 5 | 580 | 3459189 | ||
BP228371.1 | - | 3, 4 | 324 | |||
CN256547.1 | - | 5, 6 | 649 | |||
DA567999.1 | - | 4, 5 | 394 | |||
DA571733.1 | - | 5, 6 | 667 | |||
DQ020163.1 | - | 4, 5 | 552 |