Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024574.3 | NP_078850.3 | 2 | 693 | ||
GenBank mRNA | AK289615.1 | - | 2 | 601 | ||
AY358821.1 | - | 2 | 265 | |||
BC019351.1 | - | 2 | 589 | 3928768 | ||
BX359842.2 | - | 2 | 691 | |||
DA807168.1 | - | 1 | 177 |